missing translation for 'onlineSavingsMsg'
Learn More

CXCR7 Mouse, BV421, Clone: 10D1, BD Horizon™

Mouse Monoclonal Antibody

Marke:  BD Biosciences 566233

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15894859

  • 1195.00 € / 1 Stück
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

CXCR7 Monoclonal antibody specifically detects CXCR7 in Human, Mouse samples. It is validated for Flow cytometry.

CXCR7 Monoclonal antibody specifically detects CXCR7 in Human, Mouse samples. It is validated for Flow cytometry.
Spezifikation

Spezifikation

CXCR7
Monoclonal
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
0.1 mg
Cell Biology
Human, Mouse
Purified
Flow Cytometry
10D1
Brilliant Violet 421
ACKR3, CMKOR, RDC1, GPR159
Human CXCR7 Transfected Cell Line
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed.
IgG2a κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.