missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E (G1/S-Phase Cyclin) Mouse, Clone: CCNE1/2460, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 898-MSM1-P0
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Cyclin E (G1/S-Phase Cyclin) | |
Nuclear | |
CCNE1/2460 | |
Unconjugated | |
Mouse | |
53kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
898 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human Cyclin E (CCNE1) protein fragment (apprx. aa 10-176) | |
HeLa Cells. Breast or Colon Carcinoma | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |