Learn More
Cyclin H Mouse anti-Human, Unconjugated, Clone: 2C5G11, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 67065-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Cyclin H antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cyclin H. This antibody recognizes human, mouse, rat antigen. The Cyclin H antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
| Cyclin H | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 2C5G11 | |
| Unconjugated | |
| 150 μL | |
| 902 | |
| IgG2a |