Learn More
ACROBiosystems Cynomolgus CD3E&CD3D Heterodimer Protein, Llama Fc&Llama Fc, low endotoxin
low endotoxin
Marke:  ACROBiosystems CDD-C5259-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16407369
  • 447.57 € / 100 Mikrogramm
Verfügbar ab: 31-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Cynomolgus CD3E & CD3D Heterodimer Protein, Llama IgG2b Fc Tag & Llama IgG2b Fc Tag, low endotoxin is produced by co-expression of CD3E and CD3D, has a calculated MW of 41.9 kDa (CD3E) and 40.8 kDa (CD3D). Subunit CD3E is fused with a llama IgG2b Fc tag a
Spezifikation
Spezifikation
NP_001270544.1 (CD3E) and NP_001274617.1 (CD3D)
100 μg
Gln 22 - Asp 117 (CD3E) and Phe 22 - Ala 105 (CD3D)
CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon
Unconjugated
Llama IgG Fc Tag
Less than 0.01 EU / μg by the LAL method.
Powder
>90% as determined by reduced SDS-PAGE
41.9 kDa (CD3E) and 40.8 kDa (CD3D)
Cynomolgus CD3E&CD3D; Heterodimer Protein, Llama Fc&Llama; Fc, low endotoxin (CDD-C5259) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # Q95LI5-1 (CD3E) and Q95LI8-1 (CD3D)).
-20°C
CD3E and CD3D
Cynomolgus Monkey
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens