missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Cynomolgus Glypican 3 / GPC3 Protein, Fc Tag
350.80 € - 2419.31 €
Spezifikation
Zugriffsnummer | XP_005594665.1 |
---|---|
Molekulargewicht | 38.1 kDa and 49.4 kDa |
Quelle | Cynomolgus Glypican 3, Fc Tag (GP3-C5256) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # XP_005594665.1). |
Immunogen | Gln 25 - His 559 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15886867
|
ACROBiosystems
GP3-C5256-1MG |
1 mg |
2419.31 €
1 Milligramm |
Verfügbar ab: 28-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15876867
|
ACROBiosystems
GP3-C5256-100UG |
100 μg |
350.80 €
100 Mikrogramm |
Verfügbar ab: 28-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Glypican-3 (GPC3) is also known as Intestinal protein OCI-5, GTR2-2, MXR7, which belongs to the glypican family. Glypican 3 / GPC-3 is highly expressed in lung, liver and kidney. Glypican-3 inhibits the dipeptidyl peptidase activity of DPP4. Glypican-3 may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and also may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function.
This protein carries a human IgG1 Fc tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be partially processed into N and C-terminal fragment with calculated MW of 38.1 kDa and 49.4 kDa respectivelySpezifikation
XP_005594665.1 | |
Cynomolgus Glypican 3, Fc Tag (GP3-C5256) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # XP_005594665.1). | |
-20°C | |
Glypican 3 | |
Cynomolgus Monkey | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
38.1 kDa and 49.4 kDa | |
Gln 25 - His 559 | |
GPC3,OCI5,Glypican-3,GTR2-2,MXR7,DGSX,SDYS ,SGB,SGBS,SGBS1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |