Learn More
ACROBiosystems Cynomolgus IGF-I R / CD221 Protein, His Tag
Marke: ACROBiosystems IGR-C5225-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The Insulin-like Growth Factor 1 Receptor (IGF1) is also known as CD221, JTK13. and is a transmembrane receptor that is activated by IGF-1 and by the related growth factor IGF-2. It belongs to the large class of tyrosine kinase receptors. This receptor mediates the effects of IGF-1, which is a polypeptide protein hormone similar in molecular structure to insulin. IGF1R is make up of two alpha subunits and two beta subunits ,the Both the α and β subunits are synthesized from a single mRNA precursor. The precursor is then glycosylated, proteolytically cleaved, and crosslinked by cysteine bonds to form a functional transmembrane αβ chain.The α chains are located extracellularly while the β subunit spans the membrane and are responsible for intracellular signal transduction upon ligand stimulation. IGF1R have a binding site for ATP, which is used to provide the phosphates for autophosphorylation. There is a 60% homology between IGF1R and the insulin receptor. In response to ligand binding, the α chains induce the tyrosine autophosphorylation of the β chains. This event triggers a cascade of intracellular signaling that, while somewhat cell type specific, often promotes cell survival and cell proliferation.
This protein carries a polyhistidine tag at the C-terminus.This protein contains a furin convertase cleavage site, 737-RKRR-740, and will be partially processed into N (α chain) and C-terminal fragment (partial β chain) with calculated MW of 81.1 kDa andSpezifikation
XP_005560632.1 | |
100 μg | |
Glu 31 - Asn 932 | |
IGF1R,IGFR,JTK13,CD221,MGC142170,MGC142172,MGC18216 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
81.1 kDa, 23.8 kDa | |
Cynomolgus IGF-I R, His Tag (IGR-C5225) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession # G7P9I7-1). | |
-20°C | |
IGF-I R | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |