Learn More
ACROBiosystems Cynomolgus IL-17F Protein, His Tag
423.38 € - 3108.82 €
Spezifikation
Zugriffsnummer | XP_005552814.1 |
---|---|
Molekulargewicht | 16.8 kDa |
Quelle | Cynomolgus IL-17 F, His Tag (ILF-C5243) is expressed from human 293 cells (HEK293). It contains AA Arg 31 - Gln 163 (Accession # G7P4V0-1). |
Immunogen | Arg 31 - Gln 163 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
17116441
|
ACROBiosystems
ILF-C5243-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
17126441
|
ACROBiosystems
ILF-C5243-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Description
Interleukin-17F (IL17F) is also known as Interleukin-24 (IL24), Cytokine ML-1, is a secreted disulfide-linked homodimer which belongs to the IL-17 family. There are at least six members of the IL-17 family in humans and in mice: IL-17A, IL-17B, IL-17C, IL-17D, IL-17E (also called IL-25) and IL-17F (also called IL-24). IL-17 represents a family of structurally related cytokines that share a highly conserved C-terminal region but differ from one another in their N-terminal regions and in their distinct biological roles. IL-17F / IL-24 is expressed in activated, but not resting, CD4+ T-cells and activated monocytes. IL17F / Interleukin-24 stimulates the production of other cytokines such as IL-6, IL-8 and granulocyte colony-stimulating factor, and can regulate cartilage matrix turnover. IL17F / IL-24 stimulates PBMC and T-cell proliferation and Inhibits angiogenesis. Defects in IL17F are the cause of familial candidiasis type 6 (CANDF6).
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
XP_005552814.1 | |
Cynomolgus IL-17 F, His Tag (ILF-C5243) is expressed from human 293 cells (HEK293). It contains AA Arg 31 - Gln 163 (Accession # G7P4V0-1). | |
-20°C | |
IL-17 F | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |
16.8 kDa | |
Arg 31 - Gln 163 | |
IL-17 F,Interleukin-17 F,Cytokine ML-1 | |
Unconjugated | |
His Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |