missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Cynomolgus LAG-3 / CD223 Protein, Fc Tag
423.38 € - 2818.49 €
Specifications
Zugriffsnummer | XP_005570011.1 |
---|---|
Molekulargewicht | 73.0 kDa |
Quelle | Cynomolgus LAG-3, Fc Tag (LA3-C5252) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession # XP_005570011.1). |
Immunogen | Ala 18 - His 449 |
Lagerungsbedingungen | -20°C |
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Menge | Price | Quantity & Availability | |||||
15887207
|
ACROBiosystems
LA3-C5252-1MG |
1 mg |
2818.49 €
1 Milligramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15877207
|
ACROBiosystems
LA3-C5252-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Description
Lymphocyte activation gene 3 protein (LAG3) is also known as CD antigen CD223 and protein FDC, which belongs to immunoglobulin (Ig) superfamily and contains 4 extracellular Ig-like domains. The LAG3 gene contains 8 exons. The sequence data, exon/intron organization, and chromosomal localization all indicate a close relationship of LAG3 to CD4. LAG3 /CD223 involved in lymphocyte activation. LAG3 /CD223 binds to HLA class-II antigens.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.0 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
XP_005570011.1 | |
Cynomolgus LAG-3, Fc Tag (LA3-C5252) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession # XP_005570011.1). | |
-20°C | |
LAG-3 | |
Cynomolgus Monkey | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
73.0 kDa | |
Ala 18 - His 449 | |
LAG3,CD223,FDC | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |