Learn More
ACROBiosystems Cynomolgus LILRB4 / CD85k / ILT3 Protein, Mouse IgG2a Fc Tag
Marke:  ACROBiosystems LI4-C5250-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15897257
  • 467.59 € / 100 Mikrogramm
Verfügbar ab: 11-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Leukocyte immunoglobulin-like receptor subfamily B member 4 (LILRB4) is also known as CD85 antigen-like family member K (CD85K), Immunoglobulin-like transcript 3 (ILT-3), Leukocyte immunoglobulin-like receptor 5 (LIR-5), Monocyte inhibitory receptor HM18, which belongs to the leukocyte immunoglobulin-like receptor (LIR) family. LILRB4 / CD85K contains 2 Ig-like C2-type (immunoglobulin-like) domains. CD85K is detected in monocytes, macrophages, dendritic cells, lung, natural killer cells and B-cells. LILRB4 / CD85K is receptor for class I MHC antigens. CD85K recognizes a broad spectrum of HLA-A, HLA-B, HLA-C and HLA-G alleles, involved in the down-regulation of the immune response and the development of tolerance. LILRB4 interferes with TNFRSF5-signaling and NF-kappa-B up-regulation and inhibits receptor-mediated phosphorylation of cellular proteins and mobilization of intracellular calcium ions.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 53.2 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
XP_015297198.1
53.2 kDa
100 µg
Gln 22 - Glu 259
LILRB4,ILT3,LIR5,CD85K,HM18
Unconjugated
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5
HEK293
Cynomolgus
Powder
>95% as determined by SDS-PAGE
Cynomolgus LILRB4, Mouse IgG2a Fc Tag (LI4-C5250) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # XP_015297198.1).
-20°C
LILRB4
Mouse IgG Fc Tag
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins