missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Cynomolgus Mesothelin / MSLN (296-580) Protein, His Tag
MALS verified
423.38 € - 3108.82 €
Spezifikation
Molekulargewicht | 33.7 kDa |
---|---|
Quelle | Cynomolgus Mesothelin (296-580), His Tag (MSN-C52H6) is expressed from human 293 cells (HEK293). It contains AA Asp 296 - Gly 580 (Accession # A0A2K5TW94-1). |
Immunogen | Asp 296 - Gly 580 |
Lagerungsbedingungen | -20°C |
Gen-Alias | MSLN,Mesothelin,MPF |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
17186501
|
ACROBiosystems
MSN-C52H6-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 13-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
17176501
|
ACROBiosystems
MSN-C52H6-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 13-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.7 kDa. The protein migrates as 35-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
33.7 kDa | |
Asp 296 - Gly 580 | |
MSLN,Mesothelin,MPF | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
Cynomolgus Mesothelin (296-580), His Tag (MSN-C52H6) is expressed from human 293 cells (HEK293). It contains AA Asp 296 - Gly 580 (Accession # A0A2K5TW94-1). | |
-20°C | |
Mesothelin | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |