Learn More
ACROBiosystems Cynomolgus / Rhesus macaque CD47 Protein, Fc Tag
423.38 € - 2661.24 €
Spezifikation
Zugriffsnummer | NP_001253446.1 |
---|---|
Molekulargewicht | 40.5 kDa |
Quelle | Cynomolgus / Rhesus macaque CD47, Fc Tag (CD7-C5252) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Accession # F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologus. |
Immunogen | Gln 19 - Glu 141 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15856247
|
ACROBiosystems
CD7-C5252-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15866247
|
ACROBiosystems
CD7-C5252-1MG |
1 mg |
2661.24 €
1 Milligramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Leukocyte surface antigen CD47 is also known as Antigenic surface determinant protein OA3, Integrin-associated protein (IAP) and Protein MER6. CD47 contains 1 Ig-like V-type (immunoglobulin-like) domain. CD47 is very broadly distributed on normal adult tissues. CD47 has a role in both cell adhesion by acting as an adhesion receptor for THBS1 on platelets, and in the modulation of integrins and plays an important role in memory formation and synaptic plasticity in the hippocampus by similarity. CD47 is the receptor for SIRPA, binding to which prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells. CD47 Interaction with SIRPG mediates cell-cell adhesion, enhances superantigen-dependent T-cell-mediated proliferation and costimulates T-cell activation.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.5 kDa. As a result of glycosylation, the protein migrates as 55-66 kDa under reducing (R) condition, and 100-120 kDa under non-reducing (NR) condition (SDS-PSpezifikation
NP_001253446.1 | |
Cynomolgus / Rhesus macaque CD47, Fc Tag (CD7-C5252) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Accession # F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologus. | |
-20°C | |
CD47 | |
Cynomolgus Monkey, Rhesus Macaque | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
40.5 kDa | |
Gln 19 - Glu 141 | |
CD47,MER6,IAP,OA3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |