Learn More
Cytokeratin 19 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech CL594-10712-100UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Keratins are a large family of proteins that form the intermediate filament cytoskeleton of epithelial cells. Keratin expression is highly regulated, tissue specific, and varies according to cell-state. Type I keratins consist of acidic, low molecular weight proteins with MW ranging from 40 kDa (KRT19) to 64 kDa (KRT9). Type 2 keratins consist of basic or neutral, high molecular weight proteins with MW from 52 kDa (KRT8) to 67 kDa (KRT18). Keratin 19 is a type I cytokeratin. It is a biochemical marker of skin stem cells in vivo and in vitro.
Spezifikation
Cytokeratin 19 | |
Polyclonal | |
Rabbit | |
Primary | |
Human | |
IgG |
Immunofluorescence | |
CoraLite 594 | |
100 μL | |
3880 | |
Liquid |