Learn More
Biomex™ Cytomegalovirus (CMV) Seroconversion Panels
Biomex™ Cytomegalovirus (CMV) Seroconversion Panels are intended for standard testing by diagnostic manufacturers and researchers during assay development, evaluation, troubleshooting and post-marked surveillance of IgM/IgG antibody test systems and methods.
Marke: Biomex™ SCP-CMV-005
Beschreibung
- Illustrates the onset and decline of IgM and IgG Cytomegalovirus antibodies from one individual over a period of time.
- For CMV assay evaluation purposes of IgM/IgG antibody test systems.
- Serves as validation tool for diagnostic sensitivity, determination of analytical sensitivity, identification of cut-off values or to study the humoral immune response to this infection.
- Defibrinated and filtered.
- Sodium azide (0.095%) was added as preservative.
- All panel members are ready to use.
Spezifikation
| Single donor | |
| 6 x 0.5 mL Vials | |
| −20°C to -80°C | |
| Cytomegalovirus |
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.