Learn More
DiscoverX [D-Ala2, D-Leu5]-Enkephalin (DADLE)
Beschreibung
[D-Ala2, D-Leu5]-Enkephalin (DADLE) is a synthetic peptide that is a selective agonist of the delta opioid receptor, but also binds mu opioid receptors of subtype μ1. It has antinociceptive and analgesic properties via neuron protection, and in-vivo effects include reduction in arterial blood pressure and heart rate. Provided as 5mg of dry powder alongside reconstitution buffer.
Spezifikation
Spezifikation
| Produkttyp | Control Ligands |
| Menge | 5 mg |