Learn More
Daclatasvir, MedChemExpress
Click to view available options
Menge:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Packungsgröße:
10 Milligramm
100 Milligramm
5 Milligramm
50 Milligramm
Beschreibung
Daclatasvir (BMS-790052) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir is also a organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.
Specifications
Specifications
| Chemischer Name oder Material | Daclatasvir |
| Physikalische Form | Solid |
| CAS | 1009119-64-5 |
| Farbe | Light Yellow |
| Menge | 50 mg |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C40H50N8O6 |
| Zur Verwendung mit (Anwendung) | COVID-19-anti-virus |
| Synonym | BMS-790052 EBP 883 |
| Löslichkeitsinformationen | DMSO : ≥ 40 mg/mL (54.14 mM) |
| Show More |