missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva DALT Cassette 1.0 mm, 1each
Used for 1mm thick gel
Marke: Cytiva 80606727
Weitere Details : Netto-Gewicht : 1.00000kg
Beschreibung
- No assembly required
- Two glass plates are attached along one edge by a flexible silicone rubber hinge
- Spacers (1.0 or 1.5mm thick) are permanently attached to one of the glass plates
- No clamps or tape are required
- 25 x 20cm cassette produces a gel approximately 23cm wide and 19cm high
- Easily accommodates an 18cm IPG strip with additional room for molecular weightmarkers and standards
- After separation, remove gels by opening and lifting out the slab gel
- Easily cleaned and dishwasher safe
Spezifikation
Gel Cassette | |
0.03 in. | |
0.1 cm |