Learn More
Invitrogen™ DCLK1 Recombinant Rabbit Monoclonal Antibody (24GB4805)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557388
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
DCLK1 Recombinant Monoclonal Antibody for Western Blot, Flow
DCAMKL1 (DCLK1) serine/threonine kinase or DCLK1 or doublecortin-like kinase 1 contains two N-terminal doublecortin domains (which bind microtubules and regulate microtubule polymerization), a C-terminal serine/threonine protein kinase domain (which shows substantial homology to Ca2+/calmodulin-dependent protein kinase), and a serine/proline-rich domain in between the doublecortin and the protein kinase domains (which mediates multiple protein-protein interactions). DCAMKL1 is a microtubule-associated kinase that can undergo autophosphorylation. DCAMKL1 has microtubule-polymerizing activity that is independent of its protein kinase activity. DCAMKL1 is involved in several different cellular processes, including neuronal migration, retrograde transport, neuronal apoptosis and neurogenesis.
Spezifikation
| DCLK1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DCLK1 | |
| 1700113D08Rik; 2810480F11Rik; AI836758; Ania4; Calcium/calmodulin-dependent protein kinase type I-like CPG16; CL1; CLICK1; Click-I; CLICK-I beta; Cpg16; Dcamkl1; DCDC3A; Dcl; Dclk; Dclk1; double cortin and calcium/calmodulin-dependent protein kinase-like 1; doublecortin and calcium/calmodulin-dependent protein kinase-like 1; doublecortin domain-containing protein 3A; doublecortin like kinase 1; doublecortin-like and CAM kinase-like 1; Doublecortin-like kinase 1; KIAA0369; mKIAA0369; RP11-113P14.1; Serine/threonine-protein kinase DCLK1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13175, 9201 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4805 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15075, Q9JLM8 | |
| DCLK1 | |
| A synthesized peptide derived from human DCAMKL1 (aa 664-678). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |