missing translation for 'onlineSavingsMsg'
Learn More

Dengue Virus 2 premembrane (prM) Envelope Mouse anti-Virus, Clone: CC5.A9.E10, Novus Biologicals™

Mouse Monoclonal Antibody

526.00 € - 1290.00 €

Spezifikation

Antigen Dengue Virus 2 premembrane (prM) Envelope
Klon CC5.A9.E10
Konzentration 1 mg/mL
Verdünnung Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Anwendungen Western Blot, ELISA, Immunofluorescence
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
18390126
Unterlagen ansehen
Novus Biologicals
NBP3-14844-0.1MG
0.1 mg
526.00 €
0.10 Milligramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
18391276
Unterlagen ansehen
Novus Biologicals
NBP3-14844-0.5MG
0.5 mg
1290.00 €
0.50 Milligramm
View alternative products
missing translation for 'toSaveUpto' 
»
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Description

Description

Dengue Virus 2 premembrane (prM) Envelope Monoclonal antibody specifically detects Dengue Virus 2 premembrane (prM) Envelope in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications

Specifications

Dengue Virus 2 premembrane (prM) Envelope
1 mg/mL
Western Blot, ELISA, Immunofluorescence
Unconjugated
Mouse
Virus
Recombinant Dengue virus Virus-like particles, a mixture of serotypes 1-4
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
CC5.A9.E10
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Monoclonal
Purified
RUO
PBS pH 7.4
IgG1
Protein G purified
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.