missing translation for 'onlineSavingsMsg'
Learn More

Dichlorisone acetate, MedChemExpress

Artikelnummer. 16484587
Encompass_Preferred
Change view
Click to view available options
Menge:
10 mg
50 mg
Packungsgröße:
10 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16484587 50 mg -
16458698 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16484587 Lieferant Medchem Express Lieferanten-Nr. HYB1383

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

MedChemExpress Dichlorisone acetate is a synthetic glucocorticoid corticosteroid used as an anti-inflammatory agent.

Dichlorisone acetate is a synthetic glucocorticoid corticosteroid used as an anti-inflammatory agent.

Specifications

Chemischer Name oder Material Dichlorisone acetate
CAS 79-61-8
Farbe White
Hinweise zur Reinheitsqualität Research
Summenformel C23H28Cl2O5
Menge 50 mg
Löslichkeitsinformationen DMSO : 4.35 mg/mL (9.55 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@@]12[C@](C(COC(C)=O)=O)(O)CC[C@@]1([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@@]3(Cl)[C@@H](Cl)C2)=O
Molekulargewicht (g/mol) 455.37
Formelmasse 455.37
Reinheit (%) 99.77%
Güte Research
Physikalische Form Solid
Zur Verwendung mit (Anwendung) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.