Learn More
Diethylcyclopropan-1,1-Dicarboxylat, Thermo Scientific™
Click to view available options
Menge:
1g
5g
25g
1559-02-0
C9H14O4
186.21
MFCD00001283
KYYUCZOHNYSLFV-UHFFFAOYSA-N
diethyl 1,1-cyclopropanedicarboxylate, 1,1-diethyl cyclopropane-1,1-dicarboxylate, 1,1-cyclopropanedicarboxylic acid diethyl ester, 1,1-cyclopropanedicarboxylic acid, diethyl ester, cyclopropane-1,1-dicarboxylic acid diethyl ester, 1,1-bis ethoxycarbonyl cyclopropane, diethylcyclopropan-1,1-dicarboxylat, diethyl-1,1-cyclopropanedicarboxylate, acmc-209dcw, ksc495e2n
73790
Diethylcyclopropan-1,1-dicarboxylat
CCOC(=O)C1(CC1)C(=O)OCC
1559-02-0
C9H14O4
186.21
MFCD00001283
KYYUCZOHNYSLFV-UHFFFAOYSA-N
diethyl 1,1-cyclopropanedicarboxylate, 1,1-diethyl cyclopropane-1,1-dicarboxylate, 1,1-cyclopropanedicarboxylic acid diethyl ester, 1,1-cyclopropanedicarboxylic acid, diethyl ester, cyclopropane-1,1-dicarboxylic acid diethyl ester, 1,1-bis ethoxycarbonyl cyclopropane, diethylcyclopropan-1,1-dicarboxylat, diethyl-1,1-cyclopropanedicarboxylate, acmc-209dcw, ksc495e2n
73790
Diethylcyclopropan-1,1-dicarboxylat
CCOC(=O)C1(CC1)C(=O)OCC
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemischer Name oder Material Diethyl cyclopropane-1,1-dicarboxylate
Dichte 1.055
Siedepunkt 94°C to 96°C (10mmHg)
Flammpunkt 87°C (189°F)
Geruch Odorless
Beilstein 2049509
Brechungsindex 1.433
Menge 1g
Formelmasse 186.21
RUO – Research Use Only