missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, ≥99.5%, Carl Roth™

Product Code. 10032433
Encompass_Preferred
Change view
Click to view available options
Menge:
1 L
Verpackung:
Glas
Unit Size:
1 Liter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Verpackung unitSize
10032433 1 L Glas 1 Liter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10032433 Supplier Carl Roth Supplier No. 7029.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 67-68-5
Schmelzpunkt 18.5°C
Verpackung Glas
Summenformel C2H6OS
MDL-Nummer MFCD00002089
Menge 1 L
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
InChI-Schlüssel IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES CS(C)=O
IUPAC-Name Methansulfinylmethan
Molekulargewicht (g/mol) 78.13
PubChem CID 679
ChEBI CHEBI:28262
Physikalische Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.