missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA polymerase mu Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP2-57664
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
DNA polymerase mu Polyclonal specifically detects DNA polymerase mu in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Spezifikation
| DNA polymerase mu | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| DNA polymerase mu, DNA-directed DNA/RNA polymerase mu, EC 2.7.7.7, FLJ35482, Pol iota, Pol Mu, polmu, polymerase (DNA directed), mu, polymerase (DNA-directed), mu, Tdt-N, Terminal transferase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| POLM | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SEATHVVMEETSAEEAVSWQERRMAAAPPGCTPPALLDISWLTESLGAGQPVPVECRHRLEVAGPRKGPLSPAWMPAYACQRPTPLTHHNTG | |
| 100 μL | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 27434 | |
| Human | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur