Learn More
DNA-RNA Hybrid Antibody (S9.6), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-44628-20ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
DNA-RNA Hybrid Monoclonal antibody specifically detects DNA-RNA Hybrid in All samples. It is validated for ChIP assay,Flow Cytometry,Immunoprecipitation,Immunocytochemistry/ Immunofluorescence
Spezifikation
| DNA-RNA Hybrid | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| DNA-RNA heteropolymer duplex prepared by transcription of phi X174 single-stranded DNA with DNA-dependent RNA polymerase | |
| 20 μg | |
| Primary | |
| Store at 4⪚C. | |
| IgG2a κ |
| ChIP Assay, Flow Cytometry, Immunoprecipitation, Immunocytochemistry/Immunofluorescence | |
| S9.6 | |
| Unconjugated | |
| Mouse | |
| Protein A purified | |
| RUO | |
| All species | |
| Purified |