Learn More
DNMT3A / DNA Methyltransferase 3 Alpha Mouse, Clone: PCRP-DNMT3A-1E2, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 1788-MSM1-P0
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
| DNMT3A / DNA Methyltransferase 3 Alpha | |
| Nuclear | |
| PCRP-DNMT3A-1E2 | |
| Unconjugated | |
| Mouse | |
| 102kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 1788 | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human DNMT3A protein | |
| HeLa cells orPlacenta | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |