Learn More
DNMT3A / DNA Methyltransferase 3 Alpha Mouse, Clone: PCRP-DNMT3A-1E2, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 478.00 €
Spezifikation
| Antigen | DNMT3A / DNA Methyltransferase 3 Alpha |
|---|---|
| Klon | PCRP-DNMT3A-1E2 |
| Konzentration | 200μg/mL |
| Anwendungen | Immunoprecipitation, Western Blot, Immunohistochemistry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15979887
|
EnquireBio
1788-MSM1-P0 |
20 μg | |||||||
|
15989887
|
EnquireBio
1788-MSM1-P1 |
100 μg |
554.40 € 478.00 € / 100 Mikrogramm Sparen 76.40 € 13% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Spezifikation
| DNMT3A / DNA Methyltransferase 3 Alpha | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human DNMT3A protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 102kDa |
| PCRP-DNMT3A-1E2 | |
| Immunoprecipitation, Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 1788 | |
| IgG1 | |
| Protein A/G purified from Bioreactor concentrate. | |
| Nuclear | |
| HeLa cells orPlacenta |