Learn More
DNPEP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15016-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Aminopeptidase with specificity towards an acidic amino acid at the N-terminus. Likely to play an important role in intracellular protein and peptide metabolism.Spezifikation
| DNPEP | |
| Polyclonal | |
| Unconjugated | |
| DNPEP | |
| aspartyl aminopeptidase, ASPEP, DAP, DNPEP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23549 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9ULA0 | |
| DNPEP | |
| DNPEP Fusion Protein Ag6966 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |