missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DSPG3 Antibody, R&D Systems™
Goat Polyclonal Antibody
N.V.
Spezifikation
Antigen | DSPG3/EPYC |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Beschreibung
DSPG3/EPYC Polyclonal specifically detects DSPG3/EPYC in Mouse samples. It is validated for Western Blot.Spezifikation
DSPG3/EPYC | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
Dermatan sulfate proteoglycan 3dermatan sulphate proteoglycan 3, DSPG3proteoglycan-lb, epiphycan, EPYC, PG-Lb, PGLB, Proteoglycan-Lb, SLRR3B, SLRR3Bepiphycan proteoglycan, Small chondroitin/dermatan sulfate proteoglycan | |
EPYC | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P70186 | |
1833 | |
Mouse myeloma cell line NS0-derived recombinant mouse DSPG3 Ala20-Ile322 Accession # P70186 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse DSPG3 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human DSPG3 is observed. |
For Research Use Only