Learn More
Duplex 2209, 60 Mesh gewebt aus Draht mit Durchmesser von 0.15 mm (0.006 Zoll), Thermo Scientific™
Marke: Thermo Scientific Alfa Aesar 046525.FL
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Duplex 2209 | |
75 x 75 mm | |
Odorless |
60 mesh woven from 0.15mm (0.006mm) dia wire | |
60 Mesh | |
Drahtgewebe |