missing translation for 'onlineSavingsMsg'
Learn More

(E)-1-(3-(3,4,5-Trimethoxyphenyl)acryloyl)-5,6-dihydropyridin-2(1H)-one, 98%, BLD Pharm™

Product Code. 31074944
Encompass_Preferred
Change view
Click to view available options
Menge:
1 mg
5 mg
10 mg
25 mg
100 mg
250 mg
1 g
5 g
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
31074944 1 mg
31062860 5 g
31074078 10 mg
31068365 250 mg
31064805 1 g
31071565 5 mg
31057939 25 mg
31061681 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 31074944 Supplier BLD Pharm Supplier No. BD1482461mg
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material (E)-1-(3-(3,4,5-Trimethoxyphenyl)acryloyl)-5,6-dihydropyridin-2(1H)-one
CAS 20069-09-4
Summenformel C17H19NO5
MDL-Nummer MFCD00075706
Menge 1 mg
SMILES O=C1C=CCCN1C(/C=C/C2=CC(OC)=C(C(OC)=C2)OC)=O
Molekulargewicht (g/mol) 317.34
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.