Learn More
E. coli serotype K99 Pili Monoclonal Antibody (K99 12-349.2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA110796
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-10796 detects E. coli K99 pili. These antibodies do not cross-react with k99-negative E.Coli, E.Coli K88ac, E.Coli 987P, Proteus vulgaris, Pseudomonas aeruginosa, Enterbacter aerogenes, Citrobacter freundii, Klebsiella pneumoniae, or strep. agalactiae. MA1-10796 has been successfully used in ELISA applications. The MA1-10796 immunogen is purified E. coli K99 pili.
E. coli serotype K99 Pili Monoclonal antibody specifically detects E. coli serotype K99 Pili in Bacteria samples. It is validated for ELISA.Spezifikation
E. coli serotype K99 Pili | |
Monoclonal | |
1.2 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA | |
K99 12-349.2 | |
Unconjugated | |
Escherichia coli | |
Purified E. coli K99 pili. | |
500 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |