Learn More
STEMCELL Technologies EasySep™ Dead Cell Removal (Annexin V) Kit
Artikelnummer. 15902954
Encompass
Change view
Click to view available options
Menge:
1 x 109 Zellen
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15902954 1 x 109 Zellen 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15902954 Lieferant STEMCELL Technologies Lieferanten-Nr. 17899
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Immunomagnetic depletion of apoptotic (Annexin V+) cells from culture or tissue preparations
  • The EasySep™ Dead Cell Removal (Annexin V) kit is designed to deplete apoptotic (Annexin V+) cells from cell culture or tissue preparations by immunomagnetic negative selection
  • Unwanted cells are labeled with Annexin V, antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are simply poured off into a new tube
  • Annexin V binds phosphatidylserine on the outer leaflet of the cell membrane during apoptosis
Spezifikation
Zelltyp Lymphocytes
Menge 1 x 109 Zellen