Learn More
STEMCELL Technologies EasySep™ Direct Human B Cell Isolation Kit for RoboSep™
Artikelnummer. 17167951
Encompass
Click to view available options
Menge:
100 l Vollblut
Packungsgröße:
1 Set
Artikelnummer. 17167951
Marke: STEMCELL Technologies 19674RF
om dit product te kopen Registreer vandaag om een webaccount aan te maken
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • Isolate highly purified B cells directly from human whole blood without the need for density gradient centrifugation, RBC sedimentation or lysis
  • This platform provides fast and fully immunomagnetic negative selection
  • This cell isolation kit targets non-B cells for removal with EasySep™ Direct RapidSpheres™ and antibodies recognizing specific surface antigens
  • Unwanted cells are separated from desired cells using an EasySep™ magnet, and untouched target cells are then pipetted or poured into a new tube
  • ​Isolated cells can be used directly in downstream applications such as flow cytometry, culture or DNA/RNA extraction
Spezifikation
Zelltyp B Cells
Zur Verwendung mit (Anwendung) Cell Isolation
Menge 100 l Vollblut