Learn More
STEMCELL Technologies EasySep™ Direct Human CD4+ T Cell Isolation Kit
Beschreibung
- Isolate CD4+ T cells directly from human whole blood by immunomagnetic negative selection
- Desired cells undergo minimal manipulation and are untouched and highly purified
- This product labels RBCs, platelets and unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled RBCs, platelets and non-target cells are separated from the untouched CD4+ T cells in an EasySep™ magnet and poured or pipetted into a new tube
- Isolated cells are available for use in downstream uses such as flow cytometry, functional assays or culture
Spezifikation
Spezifikation
| Zelltyp | T Cells, T Cells, CD4+ |
| Zur Verwendung mit (Anwendung) | Cell Isolation |
| Menge | 100 l Vollblut |