Learn More
STEMCELL Technologies EasySep™ Human Naïve CD8+ T Cell Isolation Kit II
Click to view available options
Menge:
1 x 109 Zellen
Packungsgröße:
1 Set
Description
- The EasySep™ Human Naïve CD8+ T Cell Isolation Kit II is designed to isolate naïve CD8+ T cells from fresh peripheral blood mononuclear cells by negative selection
- The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
- The magnetically-labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
- This product replaces the EasySep™ Human Naïve CD8+ T Cell Enrichment Kit (Catalog #19158) for even faster cell isolations
Specifications
Specifications
| Zelltyp | T Cells, T Cells, CD8+ |
| Zur Verwendung mit (Anwendung) | Cell Isolation |
| Menge | 1 x 109 Zellen |