Learn More
STEMCELL Technologies EasySep™ Mouse Pan-B Cell Isolation Kit
Artikelnummer. 15551763
Encompass
Change view
Click to view available options
Menge:
1 x 109 Zellen
Artikelnummer. Menge
15551763 1 x 109 Zellen
1 options
Artikelnummer. 15551763
Marke: STEMCELL Technologies 19844
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Immunomagnetic isolation of untouched mouse pan-B cells
  • The EasySep™ Mouse Pan-B Cell Isolation Kit is designed to isolate all B cells, including conventional B-2 B cells, B-1 B cells and plasma cells from single-cell suspensions of splenocytes or other tissues by negative selection
  • Unwanted cells are targeted for removal with biotinylated antibodies directed against non pan-B cells (CD4, CD8, CD11c, CD49b, CD90.2, Ly-6C/G (Gr-1), TER119) and streptavidin-coated magnetic particles (RapidSpheres™ )
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
  • This kit is compatible for use with cells from disease models where the malignant cells (B-CLL) express CD43 or CD11b
  • For isolation of conventional B cells only, we recommend using the EasySep™ Mouse B Cell Isolation Kit (Catalog #19854)
Spezifikation
Zelltyp B Cells
Zur Verwendung mit (Anwendung) Cell Isolation
Menge 1 x 109 Zellen