missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eatons Reagenz, P2O5 ca. 7.5 % w/w

Catalog Number p-7046660
Click to view available options
Menge:
100 mL
500 mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
39394-84-8
CH4O8P2S
238.043
MFCD00070545
JHNLZOVBAQWGQU-UHFFFAOYSA-N
eaton's reagent, phosphorus pentoxide methanesulfonic acid, diphosphooxidane; methanesulfonic acid, eaton's, eaton'sreagent, eaton inverted exclamation markas reagent, eaton's reagent, p2o5 ca w/w, phosphorus pentoxide, 7.7 wt. % in methanesulfonic acid
16149098
CS(=O)(=O)O.O=P(=O)OP(=O)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
39394-84-8
CH4O8P2S
238.043
MFCD00070545
JHNLZOVBAQWGQU-UHFFFAOYSA-N
eaton's reagent, phosphorus pentoxide methanesulfonic acid, diphosphooxidane; methanesulfonic acid, eaton's, eaton'sreagent, eaton inverted exclamation markas reagent, eaton's reagent, p2o5 ca w/w, phosphorus pentoxide, 7.7 wt. % in methanesulfonic acid
16149098
CS(=O)(=O)O.O=P(=O)OP(=O)=O

Eaton's Reagent is used in the synthesis fluorine-based hole-transporting material, mono and bis-chalcone derivatives via Claisen-Schmidt condensation reaction between arylaldehydes and ketones and 4-hydroxycoumarins and 4-hydroxy-2-quinolinones derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Eaton's Reagent
Namenshinweis P2O5
Hinweise zum Prozentgehalt ≈7.5 % Wasser-in-Wasser
Strukturformel P2O5
UN-Nummer UN3265
Dichte 1.515
Löslichkeitsinformationen Decomposes in water.
Formelmasse 141.95
Brechungsindex 1.435
Menge 100 mL
Flammpunkt >110°C (230°F)
Empfindlichkeit Moisture sensitive; May form a precipitate
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'