missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Edoxaban, MedChemExpress
Click to view available options
Menge:
5 mg
10 mg
50 mg
100 mg
Packungsgröße:
10 Milligramm
5 Milligramm
50 Milligramm
Description
Edoxaban (DU-176b) is an orally active, highly potent, selective, and direct Factor Xa (FXa) inhibitor with Ki values of 0.561 and 2.98 nM for free human FXa and prothrombinase. Edoxaban exhibits more than 10,000-fold selectivity over other coagulation proteases. Edoxaban can be used in preventing thromboembolic disease research.
Specifications
Specifications
| Chemischer Name oder Material | Edoxaban |
| CAS | 480449-70-5 |
| Farbe | White |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C24H30ClN7O4S |
| Menge | 100 mg |
| Synonym | DU-176 |
| Löslichkeitsinformationen | DMSO : 10 mg/mL (18.25 mM; Need ultrasonic) |
| SMILES | O=C(N(C)C)[C@@H](CC1)C[C@H]([C@H]1NC(C(NC(C=C2)=NC=C2Cl)=O)=O)NC(C3=NC(CC4)=C(CN4C)S3)=O |
| Molekulargewicht (g/mol) | 548.06 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction