Learn More
EEF1E1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555722
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LSLLEKSLGL SKGNKYSAQG ERQIPVLQTN NGPSLTGLTT IAAHLVKQAN KEYLLGSTAE EKAIVQQWLE YRVTQVDGHS SKNDIHTLLK DL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
This gene encodes a multifunctional protein that localizes both in the cytoplasm and in the nucleus. In the cytoplasm, the encoded protein is an auxiliary component of the macromolecular aminoacyl-tRNA synthase complex. However, its mouse homolog has been shown to translocate to the nucleus in response to DNA damage and plays a positive role in ATM/ATR-mediated p53 activation. Alternative splicing results in multiple transcript variants.Spezifikation
EEF1E1 | |
Polyclonal | |
Unconjugated | |
EEF1E1 | |
1110003A02Rik; AIMP3; Aminoacyl tRNA synthetase complex-interacting multifunctional protein 3; ARS-interacting multifunctional protein 3; Eef1e1; Elongation factor p18; eukaryotic translation elongation factor 1 epsilon 1; eukaryotic translation elongation factor 1 epsilon-1; Multisynthase complex auxiliary component p18; multisynthetase complex auxiliary component p18; P18; p18 component of aminoacyl-tRNA synthetase complex | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9521 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43324 | |
EEF1E1 | |
Recombinant protein corresponding to Human EEF1E1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only