Learn More
EFS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551602
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: IYKIPRASGT QLAAPRDALE VYDVPPTALR VPSSGPYDCP ASFSHPLTRV APQPPGEDDA PYDVPLTPKP PAELEPDLEW EGGREPGPPI YAAPSNLKRA SALLNLYEAP EELLADGEGG GTDEGIYDVP LLGPEAPPSP EPPGALAS Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 78%.
Docking protein which plays a central coordinating role for tyrosine-kinase-based signaling related to cell adhesion. May serve as an activator of SRC and a downstream effector. Interacts with the SH3 domain of FYN and with CRK, SRC, and YES.Spezifikation
EFS | |
Polyclonal | |
Unconjugated | |
EFS | |
Cas scaffolding protein family member 3; CAS3; CASS3; EFS; EFS1; EFS2; embryonal Fyn-associated substrate; hEFS; signal transduction protein (SH3 containing); signal-integrating protein; Sin; SRC-interacting protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10278 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43281 | |
EFS | |
Recombinant protein corresponding to Human EFS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only