Learn More
ELSPBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560661
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SCISQGSFLG SLWWSVTSVF DEKQQWKFCE TNEYGGNSLR KPCIFPSIYR NNVVSDCMED ESNKLWCPT Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 36%.
ELSPBP1 (epididymal sperm-binding protein 1), also known as epididymal secretory protein 12 (E12), HE12 or EDDM12, is a 223 amino acid member of the seminal plasma protein family. ELSPBP1 is a secreted protein that is detected in cauda epididymal fluid and on sperm membrane. ELSPBP1 has phosphorylcholine-binding activity and has been shown to bind to spermatozoa upon ejaculation and is thought to play a role in sperm capacitation. N-glycosylated, ELSPBP1 contains four fibronectin type-II domains. The gene that encodes ELSPBP1 maps to human chromosome 19, which consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes.Spezifikation
ELSPBP1 | |
Polyclonal | |
Unconjugated | |
ELSPBP1 | |
E12; EDDM12; EL149; ELSPBP1; epididymal protein 12; epididymal secretory protein 12; epididymal sperm binding protein 1; epididymal sperm-binding protein 1; epididymis luminal secretory protein 149; ESPB1; hE12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BH3 | |
ELSPBP1 | |
Recombinant protein corresponding to Human ELSPBP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only