Learn More
EMCN Mouse, anti-Human, Unconjugated, Clone: 2D8G3, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Spezifikation
| Antigen | EMCN |
|---|---|
| Klon | 2D8G3 |
| Konzentration | 1000 μg/mL |
| Anwendungen | Western Blot, ELISA |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
17291516
|
Proteintech
67854-1-IG-20UL |
20 μL |
145.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
17281516
|
Proteintech
67854-1-IG-150UL |
150 μL |
410.00 €
150 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
The EMCN antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human EMCN. This antibody recognizes human, mouse antigen. The EMCN antibody has been validated for the following applications: WB,ELISA analysis.
Spezifikation
| EMCN | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Mouse | |
| Primary |
| 2D8G3 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| 51705 |