Learn More
EndoGlyx-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554264
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: HAQHFTLHRS ISELQADVDT KLKRLHKAQE APGTNGSLVL ATPGAGARPE PDSLQARLGQ LQRNLSELHM TTARREEELQ YTLEDMRATL TRHVDEIKEL YSESDETFDQ ISKVERQVEE LQVNHTALRE LRVILMEK Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.
Inhibits endothelial cells motility and acts as a negative regulator of angiogenesis; it downregulates KDR activation by binding VEGFA.Spezifikation
EndoGlyx-1 | |
Polyclonal | |
Unconjugated | |
MMRN2 | |
Elastin microfibril interface located protein 3; elastin microfibril interfacer 3; EMILIN3; EMILIN-3; EMILIN-like protein EndoGlyx-1; ENDOGLYX-1; endoGlyx-1 p125/p140 subunit; MMRN2; multimerin 2; multimerin-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79812 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H8L6 | |
MMRN2 | |
Recombinant protein corresponding to Human EndoGlyx-1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only