Learn More
STEMCELL Technologies EpiCult™-B Human Medium Kit
Beschreibung
- EpiCult™-B Human Medium Kit is a serum-free culture medium optimized for the culture of human mammary luminal and myoepithelial cells
- It is ideal for the growth and evaluation of bipotent, luminal-restricted and myoepithelial-restricted mammary epithelial progenitor cells in the mammary colony-forming unit (CFU) assay when used in conjunction with an irradiated feeder layer such as NIH 3T3
- This kit contains EpiCult™-B Basal Medium (Human) and EpiCult™-B Proliferation Supplement (Human)
- Addition of Hydrocortisone Stock Solution (Catalog #07925) is required
Spezifikation
Spezifikation
| Zelltyp | Mammary Cells |
| Zur Verwendung mit (Anwendung) | Cell Culture; Colony Assay |
| Menge | 1 Kit |