Learn More
Biomex™ Epstein Barr Virus (EBV) Seroconversion Panels
Description
- Illustratess the onset and decline of IgM and IgG Epstein Barr virus antibodies from one individual over a period of time.
- For EBV assay evaluation purposes of IgM/IgG antibody test systems.
- Serves as validation tool for diagnostic sensitivity, determination of analytical sensitivity, identification of cut-off values or to study the humoral immune response to this infection.
Specifications
Specifications
| Beschreibung | Single donor |
| Menge | 25 x 0.5 mL Vials |
| Inhalt und Lagerung | −20°C to -80°C |
| Produkttyp | Epstein Barr virus |