Learn More
ER alpha/NR3A1 Antibody (EVG F9), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB100-74413-0.025ml
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human, Mouse, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Spezifikation
| ER alpha/NR3A1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.05% Sodium Azide | |
| ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
| Mouse | |
| 67 kDa | |
| 0.025 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| 2099 | |
| Human | |
| Ascites |
| Western Blot | |
| EVG F9 | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200 | |
| P03372 | |
| ESR1 | |
| Mix of synthetic peptides corresponding to residuesT(140) V R E A G P P A F Y R P N S(154) & E(247) V G M M K G G I R K D R R G G R(263) of human ER alpha. | |
| Unpurified | |
| RUO | |
| Primary | |
| Estrogen Receptor alpha (EVG F9) | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |