missing translation for 'onlineSavingsMsg'
Learn More

(Ethoxycarbonylmethyl)dimethylsulfoniumbromid, 98 %, Thermo Scientific Chemicals

Catalog Number p-7043330
Click to view available options
Menge:
10 g
50 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5187-82-6
C6H13BrO2S
229.132
MFCD00830557
JXFPTJYKYKVENJ-UHFFFAOYSA-M
ethoxycarbonylmethyl dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfonium bromide, carbethoxymethyl-dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfanium bromide, 2-ethoxy-2-oxoethyl dimethyl sulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulphonium bromide, sulfonium, carbethoxymethyl dimethyl-, bromide, sulfonium, 2-ethoxy-2-oxoethyl dimethyl-, bromide, carbethoxymethyl dimethylsulfonium bromide, acmc-1az78
78861
(2-Ethoxy-2-Oxoethyl)-Dimethylsulfanium;Bromid
CCOC(=O)C[S+](C)C.[Br-]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5187-82-6
C6H13BrO2S
229.132
MFCD00830557
JXFPTJYKYKVENJ-UHFFFAOYSA-M
ethoxycarbonylmethyl dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfonium bromide, carbethoxymethyl-dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfanium bromide, 2-ethoxy-2-oxoethyl dimethyl sulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulphonium bromide, sulfonium, carbethoxymethyl dimethyl-, bromide, sulfonium, 2-ethoxy-2-oxoethyl dimethyl-, bromide, carbethoxymethyl dimethylsulfonium bromide, acmc-1az78
78861
(2-Ethoxy-2-Oxoethyl)-Dimethylsulfanium;Bromid
CCOC(=O)C[S+](C)C.[Br-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (Ethoxycarbonylmethyl)dimethylsulfonium bromide
Schmelzpunkt ∽80°C (decomposition)
Menge 10 g
Beilstein 4161472
Empfindlichkeit Hygroscopic
Formelmasse 229.14
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'