missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-4-chlor-6,8-dimethoxychinolin-3-carboxylat, 97 %, Thermo Scientific™

Catalog Number p-7039661
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1016825-43-6
C14H14ClNO4
295.719
MFCD09937855
DSGVVCRDDXQCTD-UHFFFAOYSA-N
4-Chloro-6,8-dimethoxyquinoline-3-carboxylic acid ethyl ester
24698979
Ethyl 4-Chlor-6,8-Dimethoxychinolin-3-Carboxylat
CCOC(=O)C1=C(C2=CC(=CC(=C2N=C1)OC)OC)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1016825-43-6
C14H14ClNO4
295.719
MFCD09937855
DSGVVCRDDXQCTD-UHFFFAOYSA-N
4-Chloro-6,8-dimethoxyquinoline-3-carboxylic acid ethyl ester
24698979
Ethyl 4-Chlor-6,8-Dimethoxychinolin-3-Carboxylat
CCOC(=O)C1=C(C2=CC(=CC(=C2N=C1)OC)OC)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 4-chloro-6,8-dimethoxyquinoline-3-carboxylate
Menge 250 mg
Formelmasse 295.72
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'