missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-4-(maleimidomethyl)-2-(2-pyridyl)-thiazol-5-carboxylat, 97 %, Thermo Scientific™

Catalog Number p-7039561
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1858256-22-0
C16H13N3O4S
343.357
MFCD22988977
LSOCKOVBJWMGKX-UHFFFAOYSA-N
ethyl 4-2,5-dioxopyrrol-1-yl methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylate, ethyl 4-maleimidomethyl-2-2-pyridyl thiazole-5-carboxylate
98000538
Ethyl 4-[(2,5-Dioxopyrrol-1-yl)Methyl]-2-Pyridin-2-yl-1,3-Thiazol-5-Carboxylat
CCOC(=O)C1=C(N=C(S1)C2=CC=CC=N2)CN3C(=O)C=CC3=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1858256-22-0
C16H13N3O4S
343.357
MFCD22988977
LSOCKOVBJWMGKX-UHFFFAOYSA-N
ethyl 4-2,5-dioxopyrrol-1-yl methyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylate, ethyl 4-maleimidomethyl-2-2-pyridyl thiazole-5-carboxylate
98000538
Ethyl 4-[(2,5-Dioxopyrrol-1-yl)Methyl]-2-Pyridin-2-yl-1,3-Thiazol-5-Carboxylat
CCOC(=O)C1=C(N=C(S1)C2=CC=CC=N2)CN3C(=O)C=CC3=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl 4-(maleimidomethyl)-2-(2-pyridyl)thiazole-5-carboxylate
Menge 1 g
Formelmasse 343.36
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'