missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 9-bromononanoate, 97%, BLD Pharm™

Artikelnummer. 30967784
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
100 mg
250 mg
1 g
5 g
10 g
25 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30967784 250 mg
30963269 25 g
30964990 1 g
30953514 10 g
30962340 100 mg
30950309 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Dieser Artikel kann nicht zurückgegeben werden. View return policy
Product Code. 30967784 Supplier BLD Pharm Supplier No. BD13493250mg
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material Ethyl 9-bromononanoate
CAS 28598-81-4
Summenformel C11H21BrO2
MDL-Nummer MFCD06797065
Menge 250 mg
SMILES O=C(OCC)CCCCCCCCBr
Molekulargewicht (g/mol) 265.19
Reinheit (%) 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.