missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-(-)-3-Hydroxybutyrat

Catalog Number p-7042693
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
24915-95-5
C6H12O3
132.159
MFCD00075386
OMSUIQOIVADKIM-RXMQYKEDSA-N
ethyl r-3-hydroxybutyrate, ethyl 3r-3-hydroxybutanoate, ethyl r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, r-ethyl 3-hydroxybutanoate, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, butanoic acid, 3-hydroxy-, ethyl ester, 3r, r---3-hydroxy-n-butyric acid ethyl ester
440030
CHEBI:28707
Ethyl-(3R)-3-hydroxybutanoat
CCOC(=O)CC(C)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
24915-95-5
C6H12O3
132.159
MFCD00075386
OMSUIQOIVADKIM-RXMQYKEDSA-N
ethyl r-3-hydroxybutyrate, ethyl 3r-3-hydroxybutanoate, ethyl r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, r-ethyl 3-hydroxybutanoate, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, butanoic acid, 3-hydroxy-, ethyl ester, 3r, r---3-hydroxy-n-butyric acid ethyl ester
440030
CHEBI:28707
Ethyl-(3R)-3-hydroxybutanoat
CCOC(=O)CC(C)O

Zur Synthese optisch aktiver Produkte

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Ethyl (R)-(-)-3-hydroxybutyrate
Farbe Farblos
Menge 1 g
Formelmasse 132.16
Physikalische Form Flüssig
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'